EMAIL THIS PAGE TO A FRIEND
The Canadian veterinary journal = La revue veterinaire canadienne
Compounding errors in 2 dogs receiving anticonvulsants.
PMID 23024385
Abstract
Two cases that involve drug compounding errors are described. One dog exhibited increased seizure activity due to a compounded, flavored phenobarbital solution that deteriorated before the expiration date provided by the compounder. The other dog developed clinical signs of hyperkalemia and bromine toxicity following a 5-fold compounding error in the concentration of potassium bromide (KBr).