EMAIL THIS PAGE TO A FRIEND
Journal of the Indian Medical Association
Corneal ulcer with Scopulariopsis brevicaulis and Staphylococcus aureus--a rare case report.
PMID 23025228
Abstract
Scopulariopsis brevicaulis is a saprophytic fungus, found in soil. It causes human nail infection. Corneal ulcer with this organism is unusual. Here a case of a 36-year-old farmer is reported who developed a paracentral corneal ulcer in the right eye. The ulcer was caused by mixed infection with Staphylococcus aureus and Scopulariopsis brevicaulis. It was treated with topical natamycin 5% and moxifloxacin 0.5% drops. This is a unique case of corneal ulcer with mixed infection of Staphylococcus aureus and Scopulariopsis brevicaulis without any history of trauma, which was treated successfully, resulting in a minimal corneal opacity.