EMAIL THIS PAGE TO A FRIEND
Optics letters
Tm:LiYF4 planar waveguide laser at 1.9 μm.
PMID 23027269
Abstract
Continuous wave laser operation at 1.87 μm of liquid-phase epitaxially (LPE) grown Tm(3+)-doped YLiF(4) (Tm:YLF) layers is demonstrated. The waveguide laser delivers 560 mW by pumping with a Ti:Sapphire laser at 780 nm leading to an efficiency of 76% with respect to the absorbed pump power. This constitutes the first Tm(3+)-doped crystalline fluoride waveguide laser ever demonstrated as well as a record in efficiency and output power for an LPE grown waveguide laser operating in the 2 μm spectral range.
Related Materials