EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section F, Structural biology and crystallization communications
Structure of β-1,4-mannanase from the common sea hare Aplysia kurodai at 1.05 Å resolution.
PMID 23027740
Abstract
β-1,4-Mannanase (EC 3.2.1.78) catalyzes the hydrolysis of β-1,4-glycosidic bonds within mannan, a major constituent group of the hemicelluloses. Bivalves and gastropods possess β-1,4-mannanase and may degrade mannan in seaweed and/or phytoplankton to obtain carbon and energy using the secreted enzymes in their digestive systems. In the present study, the crystal structure of AkMan, a gastropod β-1,4-mannanase prepared from the common sea hare Aplysia kurodai, was determined at 1.05 Å resolution. This is the first report of the three-dimensional structure of a gastropod β-1,4-mannanase. The structure was compared with bivalve β-1,4-mannanase and the roles of residues in the catalytic cleft were investigated. No obvious binding residue was found in subsite +1 and the substrate-binding site was exposed to the molecular surface, which may account for the enzymatic properties of mannanases that can digest complex substrates such as glucomannan and branched mannan.
Related Materials