EMAIL THIS PAGE TO A FRIEND
Physical review letters
Self-similar gold-nanoparticle antennas for a cascaded enhancement of the optical field.
PMID 23031130
Abstract
We experimentally demonstrate cascaded field enhancement by means of gold nanoparticle dimer and trimer antennas. The local field enhancement is probed by single-molecule fluorescence using fluorophores with high intrinsic quantum efficiency (Q(0)>80%). Using a self-similar trimer antenna consisting of 80, 40, and 20 nm gold nanoparticles, we demonstrate a fluorescence enhancement of 40 and a spatial confinement of 15 nm. Compared with a single gold nanoparticle, the self-similar trimer antenna improves the enhancement-confinement ratio by more than an order of magnitude. Self-similar antennas hold promise for high-resolution imaging and spectroscopy, ultrasensitive detection, and efficient single-photon sources.