EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Site-specific functionalization of RNA molecules by an unnatural base pair transcription system via click chemistry.
PMID 23032097
Abstract
Toward new biotechnology by genetic alphabet expansion, we developed an efficient site-specific labeling method for large RNA molecules. The combination of unnatural base pair transcription and post-transcriptional modification by click chemistry enables simple RNA labeling with a wide variety of functional groups at desired positions, in a one-pot reaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P73404
Pyrrole-2-carboxaldehyde, 98%
C5H5NO