EMAIL THIS PAGE TO A FRIEND
JAMA
Mexiletine for treatment of myotonia: a trial triumph for rare disease networks.
PMID 23032555
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
462047
1-(2,6-Dimethylphenoxy)-2-propanamine hydrochloride, 97%
C11H17NO · HCl
M2727
Mexiletine hydrochloride, powder
C11H17NO · HCl