EMAIL THIS PAGE TO A FRIEND
The international journal of biochemistry & cell biology
Phospholipase D activation mediates cobalamin-induced downregulation of Multidrug Resistance-1 gene and increase in sensitivity to vinblastine in HepG2 cells.
PMID 23032700
Abstract
Failure of cancer chemotherapy due to multidrug resistance is often associated with altered Multidrug Resistance-1 gene expression. Cobalamin is the cofactor of methionine synthase, a key enzyme of the methionine cycle which synthesizes methionine, the precursor of cell S-adenosyl-methionine synthesis. We previously showed that cobalamin was able to down-regulate Multidrug Resistance-1 gene expression. Herein we report that this effect occurs through cobalamin-activation of phospholipase D activity in HepG2 cells. Cobalamin-induced down-regulation of Multidrug Resistance-1 gene expression was similar to that induced by the phospholipase D activator oleic acid and was negatively modulated by the phospholipase D inhibitor n-butanol. Cobalamin increased cell S-adenosyl-methionine content, which is the substrate for phosphatidylethanolamine-methyltransferase-dependent phosphatidylcholine production. We showed that cobalamin-induced increase in cell phosphatidylcholine production was phosphatidylethanolamine-methyltransferase-dependent. Oleic acid-dependent activation of phospholipase D was accompanied by an increased sensitivity to vinblastine of HepG2 cells while n-butanol enhanced the resistance of the cells to vinblastine. These data indicate that cobalamin mediates down-regulation of Multidrug Resistance-1 gene expression through increased S-adenosyl-methionine and phosphatidylcholine productions and phospholipase D activation. This points out phospholipase D as a potential target to down-regulate Multidrug Resistance-1 gene expression for improving chemotherapy efficacy.