EMAIL THIS PAGE TO A FRIEND
The Analyst
A terbium(III)-organic framework for highly selective sensing of cytidine triphosphate.
PMID 23032999
Abstract
Highly selective sensing of cytidine triphosphate (CTP) against other triphosphate nucleosides including ATP, GTP and UTP is successfully achieved with a luminescent terbium(III)-organic framework (TbOF) of [Tb(2)(2,3-pzdc)(2)(ox)(H(2)O)(2)](n) (2,3-pzdc(2-) = 2,3-pyrazinedicarboxylate, ox(2-) = oxalate).