EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
SPOC1 modulates DNA repair by regulating key determinants of chromatin compaction and DNA damage response.
PMID 23034801
Abstract
Survival time-associated plant homeodomain (PHD) finger protein in Ovarian Cancer 1 (SPOC1, also known as PHF13) is known to modulate chromatin structure and is essential for testicular stem-cell differentiation. Here we show that SPOC1 is recruited to DNA double-strand breaks (DSBs) in an ATM-dependent manner. Moreover, SPOC1 localizes at endogenous repair foci, including OPT domains and accumulates at large DSB repair foci characteristic for delayed repair at heterochromatic sites. SPOC1 depletion enhances the kinetics of ionizing radiation-induced foci (IRIF) formation after γ-irradiation (γ-IR), non-homologous end-joining (NHEJ) repair activity, and cellular radioresistance, but impairs homologous recombination (HR) repair. Conversely, SPOC1 overexpression delays IRIF formation and γH2AX expansion, reduces NHEJ repair activity and enhances cellular radiosensitivity. SPOC1 mediates dose-dependent changes in chromatin association of DNA compaction factors KAP-1, HP1-α and H3K9 methyltransferases (KMT) GLP, G9A and SETDB1. In addition, SPOC1 interacts with KAP-1 and H3K9 KMTs, inhibits KAP-1 phosphorylation and enhances H3K9 trimethylation. These findings provide the first evidence for a function of SPOC1 in DNA damage response (DDR) and repair. SPOC1 acts as a modulator of repair kinetics and choice of pathways. This involves its dose-dependent effects on DNA damage sensors, repair mediators and key regulators of chromatin structure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-689
Anti-HP1α Antibody, clone15.19s2, Anti-HP1α Antibody, clone15.19s2 is a Mouse Monoclonal Antibody for detection of HP1α also known as HP1Hs alpha, Heterochromatin protein 1 homolog alpha & has been validated in ChIP, ICC, IHC, IP & WB.
05-636
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, Anti-phospho-Histone H2A.X (Ser139), clone JBW301 is a well published Mouse Monoclonal Antibody validated in ChIP, ICC, IF, WB. This purified mAb is highly specific for phospho-Histone H2A.X (Ser139) also known as H2AXS139p.
07-550
Anti-SUV39H1 Antibody, Use Anti-SUV39H1 Antibody (Rabbit Polyclonal Antibody) validated in WB to detect SUV39H1 also known as KMT1A, Histone H3-K9 Methyltransferase 1.
05-615
Anti-SUV39H1 Antibody, clone MG44, Anti-SUV39H1 Antibody, clone MG44 is a Mouse Monoclonal Antibody for detection of SUV39H1 also known as KMT1A, Histone H3-K9 Methyltransferase 1 & has been validated in ChIP, IP & WB.
MAB3662
Anti-TIF1 beta Antibody, Anti-TIF1 beta Antibody is a Mouse Monoclonal Antibody for detection of Transcriptional Intermediary Factor 1 also known as TIF1 beta & has been validated in ELISA, WB, ICC.
07-442
Anti-trimethyl-Histone H3 (Lys9) Antibody, Use Anti-trimethyl-Histone H3 (Lys9) Antibody (rabbit polyclonal antibody) validated in DB, ICC, Mplex, PIA, WB, ChIP-seq to detect trimethyl-Histone H3 (Lys9) also known as H3K9me3, Histone H3 (tri methyl K9).