EMAIL THIS PAGE TO A FRIEND
Prikladnaia biokhimiia i mikrobiologiia
Microbial activity in the landfill soil.
PMID 23035574
Abstract
The research objective was to determine the activity of microorganisms in the soil exposed to direct influence of a landfill, as well as in the soil beyond its influence. Fluorescein diacetate (FDA) hydrolytic activity and respiration in the soil were determined. The highest number of cultivated bacteria was recorded at the site located within the zone of direct influence exerted by the landfill, whereas the least amount was found at a distance of 1000 metres from the landfill. In contrast, the largest numbers of molds were observed in the soil at a distance of 1000 m from the headquarters of the landfill. The highest FDA hydrolytic activity and biological oxygen demand (BOD5) were recorded in the soil by the headquarters of the landfill, and the least parameters were revealed at a distance of 1000 m from the landfill. It was found a high correlation between the number of bacteria and FDA hydrolytic activity of soil and BOD5 in the north-eastern of the landfill. However, in the same place, there is a low correlation between the number of molds, and FDA hydrolytic activity of soil and BOD5.