EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Silicon nanowire biosensor and its applications in disease diagnostics: a review.
PMID 23036462
Abstract
Over the past decade, silicon nanowire (SiNW) biosensors have been studied for the detection of biological molecules as highly sensitive, label-free, and electrical tools. Herein we present a comprehensive review about the fabrication of SiNW biosensors and their applications in disease diagnostics. We discuss the detection of important biomarkers related to diseases including cancer, cardiovascular diseases, and infectious diseases. SiNW biosensors hold great promise to realize point-of-care (POC) devices for disease diagnostics with potential for miniaturization and integration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
343250
Silicon, pieces, 99.95% trace metals basis
Si
267414
Silicon, powder, −60 mesh, 99.999% trace metals basis
Si
215619
Silicon, powder, −325 mesh, 99% trace metals basis
Si
633097
Silicon, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis
Si
646687
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Si
647535
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Si
647780
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Si
647802
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 3 in. × 0.5 mm
Si
647675
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm
Si
647764
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Si
647101
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Si
647543
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Si
647799
Silicon, wafer (single side polished), contains phosphorus as dopant, <111>, N-type, diam. × thickness 2 in. × 0.5 mm
Si
647705
Silicon, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Si
647772
Silicon, wafer (single side polished), <111>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Si
767492
Silicon, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.999% trace metals basis
Si
GF37993595
Silicon, rod, 100mm, diameter 10.0mm, single crystal - random orientation, 100%
Si
GF64686998
Silicon, rod, 100mm, diameter 2.0mm, crystalline, 100%
Si
GF22929307
Silicon, rod, 100mm, diameter 5.0mm, crystalline, 100%
Si
GF16363391
Silicon, rod, 10mm, diameter 2.0mm, crystalline, 100%
Si
GF66465276
Silicon, rod, 25mm, diameter 2.0mm, crystalline, 100%
Si
GF43555664
Silicon, rod, 25mm, diameter 3.15mm, single crystal - random orientation, 100%
Si
GF70438518
Silicon, rod, 40mm, diameter 20mm, single crystal, -100, 99.999%
Si
GF35274113
Silicon, rod, 40mm, diameter 20mm, single crystal, -111, 99.999%
Si
GF05482852
Silicon, rod, 50mm, diameter 2.0mm, crystalline, 100%
Si
GF91515034
Silicon, rod, 50mm, diameter 3.15mm, single crystal - random orientation, 100%
Si
GF14076930
Silicon, rod, 50mm, diameter 5.0mm, crystalline, 100%
Si
GF28448691
Silicon, rod, 50mm, diameter 5mm, single crystal, -100, 99.999%
Si
GF40945754
Silicon, rod, 50mm, diameter 5mm, single crystal, -111, 99.999%
Si
GF64342188
Silicon, rod, 50mm, diameter 6mm, single crystal, -111, 99.999%
Si