EMAIL THIS PAGE TO A FRIEND
Optics express
Quasi-nonvolatile storage in Ru-doped Bi12SiO20 crystals by two-wavelength holography.
PMID 23037015
Abstract
Prolonged read-out process of a hologram recorded at near infrared with simultaneous green light exposure is measured in Ru-doped Bi12SiO20 crystal. The experimental results are confirmed by numerical simulations, suggesting two different traps involved in the space-charge transport mechanism. In addition, quasi-permanent holographic recording of image with fast updating speed by using two-wavelength recording is demonstrated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
276332
Rubidium, ingot, 99.6% trace metals basis
Rb