EMAIL THIS PAGE TO A FRIEND
Optics express
Regrowth-free high-gain InGaAsP/InP active-passive platform via ion implantation.
PMID 23037047
Abstract
We demonstrate a regrowth-free material platform to create monolithic InGaAsP/InP photonic integrated circuits (PICs) with high-gain active and low-loss passive sections via a PL detuning of >135 nm. We show 2.5 µm wide by 400 µm long semiconductor optical amplifiers with >40 dB/mm gain at 1570 nm, and passive waveguide losses <2.3 dB/mm. The bandgap in the passive section is detuned using low-energy 190 keV channelized phosphorous implantation and subsequent rapid thermal annealing to achieve impurity-induced quantum well intermixing (QWI). The PL wavelengths in the active and passive sections are 1553 and 1417 nm, respectively. Lasing wavelengths for 500 µm Fabry-Perot lasers are 1567 and 1453 nm, respectively.