EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Production of p-xylene from biomass by catalytic fast pyrolysis using ZSM-5 catalysts with reduced pore openings.
PMID 23038098
Abstract
Pores for thought: Chemical liquid deposition of silica onto ZSM-5 catalysts led to smaller pore openings that resulted in >90% selectivity for p-xylene over the other xylenes in the catalytic fast pyrolysis of furan and 2-methylfuran (see scheme). The p-xylene selectivity increased from 51% with gallium spray-dried ZSM-5 to 72% with a pore-mouth-modified catalyst in the pyrolysis of pine wood.