EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Using smell to triage samples in point-of-care assays.
PMID 23038662
Abstract
Smell of success: Reagent 1 provides the dual readouts of odor (ethanethiol) and fluorescence (derivative of 7-hydroxycoumarin) and can be used in down-selection assays based on smell and quantitative fluorescence assays of the samples that give a positive result. An important feature of 1 is the matched sensitivity of the two outputs. This reagent is designed for use in resource-limited settings and is demonstrated in assays that detect enzymes.