EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Catalytic enantioselective cross-couplings of secondary alkyl electrophiles with secondary alkylmetal nucleophiles: Negishi reactions of racemic benzylic bromides with achiral alkylzinc reagents.
PMID 23039358
Abstract
We have developed a nickel-catalyzed method for the asymmetric cross-coupling of secondary electrophiles with secondary nucleophiles, specifically, stereoconvergent Negishi reactions of racemic benzylic bromides with achiral cycloalkylzinc reagents. In contrast to most previous studies of enantioselective Negishi cross-couplings, tridentate pybox ligands are ineffective in this process; however, a new, readily available bidentate isoquinoline-oxazoline ligand furnishes excellent ee's and good yields. The use of acyclic alkylzinc reagents as coupling partners led to the discovery of a highly unusual isomerization that generates a significant quantity of a branched cross-coupling product from an unbranched nucleophile.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I28208
Isoquinoline, 97%
C9H7N
282162
Isoquinoline, technical grade, 90-92%
C9H7N