EMAIL THIS PAGE TO A FRIEND
Euro surveillance : bulletin Europeen sur les maladies transmissibles = European communicable disease bulletin
Usefulness of the European Epidemic Intelligence Information System in the management of an outbreak of listeriosis, Belgium, 2011.
PMID 23040965
Abstract
A cluster of time-linked cases and the identification of a clonal strain suggest the occurrence of an outbreak of listeriosis in Belgium in 2011, presumably due to the consumption of hard cheese made with pasteurised milk and produced by a Belgium manufacturer. The outbreak clone was identified as Listeria monocytogenes serovar 1/2a, sensitive to arsenic and cadmium and of multilocus sequence typing MLST-type 37. Food investigation of this outbreak was facilitated by the European Epidemic Intelligence Information System and data exchanged between French and Belgium listeriosis surveillance systems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
28811
Cadmium chloride 0.1 M solution