EMAIL THIS PAGE TO A FRIEND
International journal of systematic and evolutionary microbiology
Pleomorphobacterium xiamenense gen. nov., sp. nov., a moderate thermophile isolated from a terrestrial hot spring.
PMID 23041633
Abstract
An aerobic, motile, moderately thermophilic rod, designated strain CLW(T), was isolated from a terrestrial hot spring in an exposition garden in Xiamen City, Fujian Province, the People's Republic of China. Strain CLW(T) formed beige, dry colonies on solid 2216E medium and flocks in liquid medium. Cells were Gram-stain-negative, short rods (1.0-3.0 µm long and 0.4-0.6 µm wide) with six or more polar flagella. The temperature and pH for growth of strain CLW(T) were 28-65 °C (optimum, 50-58 °C) and pH 5.5-9.5 (optimum, pH 6.0-8.0). Growth occurred in the presence of 0.3-6.0 % NaCl (optimum 2.5-4.5 %). Phylogenetic analysis based on 16S rRNA gene sequences showed that the closest relative of the isolate was Amaricoccus kaplicensis Ben 101(T) (94.3 % sequence similarity). The DNA G+C content of strain CLW(T) was 72.2 mol%. The respiratory quinone was ubiquinone 10. The predominant polar lipids consisted of phosphatidylglycerol and phosphatidylethanolamine. The major fatty acids (>10 %) were summed feature 8 (consisting of C18 : 1ω7c and/or C18 : 1ω6c), C18 : 1ω7c 11-methyl and C18 : 0. Based on phylogenetic, physiological and biochemical data and DNA G+C content, strain CLW(T) is considered to represent a novel species of a new genus in the family Rhodobacteraceae, for which the name Pleomorphobacterium xiamenense gen. nov., sp. nov. is proposed. The type strain of the type species is CLW(T) ( = LMG 26245(T) = CGMCC 1.10808(T) = MCCC 1A06272(T)).