EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Nanoaggregates of a pentacenequinone derivative as reactors for the preparation of palladium nanoparticles.
PMID 23042509
Abstract
Fluorescent nanoaggregates of pentacenequinone derivative serve as reactors for the preparation of palladium nanoparticles in aqueous medium. Further, the combination of catalytic quantities of palladium nanoparticles with NaBH(4) smoothly reduces 4-nitrophenol to 4-aminophenol in aqueous medium.