EMAIL THIS PAGE TO A FRIEND
Arteriosclerosis, thrombosis, and vascular biology
Novel mechanism of aortic aneurysm development in mice associated with smoking and leukocytes.
PMID 23042818
Abstract
The purpose of this study was to evaluate potential mechanisms promoting abdominal aortic aneurysm development with tobacco smoke (TS) exposure. Experiments used the elastase perfusion model of abdominal aortic aneurysms with smoke-free controls. The effect of TS exposure was evaluated in C57/Bl6 mice, after broad-spectrum matrix metalloproteinase inhibition with doxycycline and in mice deficient in matrix metalloproteinase-9, matrix metalloproteinase-12, Cathepsin-S, and Neutrophil Elastase. Preparations of washed marrow, spleen, and peripheral blood leukocytes were transferred to smoke-free mice from 6-week TS-exposed mice or smoke-free mice. All mice were euthanized 14 days after elastase perfusion, and the percentage of change in aortic diameter (%Δ aortic diameter) was calculated. Electron microscopy of aortic tissue from animals exposed to TS without elastase exposure did not demonstrate any ultrastructural changes. Neither doxycycline nor any specific elastase deficiency was effective at preventing an increase in %Δ aortic diameter in TS-exposed animals. Smoke exposure for 6 weeks increased the %Δ aortic diameter after a smoke-free interval of up to 6 weeks before elastase perfusion. Leukocyte preparations from TS-exposed mice localized to abdominal aortic aneurysms and increased the %Δ aortic diameter in smoke-free mice. The effect of TS on the development of abdominal aortic aneurysms is not dependent on the activity of elastolytic enzymes and persists for long periods despite cessation of TS. Alterations in leukocyte response to aortic injury appear to mediate this effect.
Related Materials