EMAIL THIS PAGE TO A FRIEND
Asian Pacific journal of tropical medicine
Clinical effects of psychological intervention and drug therapy against peptic ulcer.
PMID 23043926
Abstract
To evaluate the clinical effects of psychological interventions and drug therapy against peptic ulcer. 96 patients with peptic ulcer were divided into control group with Tagamet 800 mg per evening p.o. and trial group with psychological intervention on the basis of drug treatment. There were significant differences between the two groups (P<0.05), the trial group showed that the anxiety and depression cases declined obviously and effective rate of ulcer therapy was much higher than control group. In sum, psychological intervention combined with drug therapy provides an effective method for ulcer treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1089
Cimetidine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C10H16N6S · HCl