EMAIL THIS PAGE TO A FRIEND
Behavioural brain research
Behavioral and cortical EEG evaluations confirm the roles of both CCKA and CCKB receptors in mouse CCK-induced anxiety.
PMID 23043971
Abstract
This study investigated the roles of cholecystokinin (CCK)(A) and CCK(B) receptors on CCK-4-induced anxiety-like behaviors in mice through behavioral and neural evaluations. Anxiety-like behaviors in mice were induced by an intracerebroventricular (i.c.v.) administration of CCK-4, which can bind to both CCK(A) and CCK(B) receptors. The effects of CCK(A) and CCK(B) receptor antagonists (devazepide and CI-988, respectively) were examined using mouse anxiety tests (elevated-plus maze and light-dark box) and also by examining neuronal activities through EEG monitoring and c-Fos immunohistochemistry in the cortex and amygdala. CCK-4 (3 μg/kg of body weight i.c.v.) significantly induced mouse anxiety-like behaviors in the anxiety tests and also affected their EEG patterns with respect to pre-drug tracing, resulting in increase in spectral power in relative power distribution in the delta and theta bands (0.5-5 Hz frequency bands) and also in increase in c-Fos immunopositive neuron counts. These CCK-4 effects were completely suppressed by 1.0mg/kg CCK(B) receptor antagonist, CI-988, while the same amount of CCK(A) receptor antagonist, devazepide was partly able to suppress the same effects. These findings indicated that not only CCK(B) receptors but also CCK(A) receptors in the brain play important roles in regulating anxiety-like behaviors in mice. The present study also proposed a possibility that cortical EEG is useful for assessing anxiety.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T6515
Cholecystokinin Fragment 30-33 Amide, ≥95% (HPLC)
C29H36N6O6S
D3821
Devazepide, ≥98% (HPLC), powder
C25H20N4O2