EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
An enzyme-responsive nanocontainer as an intelligent signal-amplification platform for a multiple proteases assay.
PMID 23044599
Abstract
Based on selective pore-opening in the presence of protease, we have developed a novel signal amplification assay for multiple proteases detection and their inhibition using protein-capped mesoporous scaffolding as the substrate.