EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Metal-free catalyzed oxidative trimerization of indoles by using TEMPO in air: a biomimetic approach to 2-(1H-indol-3-yl)-2,3'-biindolin-3-ones.
PMID 23044781
Abstract
A simple, convenient and efficient metal-free catalyzed oxidative trimeric reaction of indoles toward a variety of 2-(1H-indol-3-yl)-2,3'-biindolin-3-one derivatives in moderate to excellent yields has been developed. This transformation proceeds via a tandem oxidative homocoupling reaction by using TEMPO in air as an environmentally benign oxidant. This methodology provides an alternative approach for the direct generation of all-carbon quaternary centers at the C3 position of indoles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I5605
Indoline, ReagentPlus®, 99%
C8H9N