EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and antituberculosis activity of novel unfolded and macrocyclic derivatives of ent-kaurane steviol.
PMID 23046964
Abstract
New derivatives of steviol 1, the aglycone of the glycosides of Stevia rebaudiana, including a novel class of semisynthetic diterpenoids, namely macrocyclic ent-kauranes were synthesized. These compounds possess antituberculosis activity inhibiting the in vitro growth of Mycobacterium Tuberculosis (H37R(V) strain) with MIC 5-20 μg/ml that is close to MIC 1 μg/ml demonstrated by antituberculosis drug isoniazid in control experiment. For the first time it was found that the change of ent-kaurane geometry (as in steviol 1) of tetracyclic diterpenoid skeleton to ent-beyerane one (as in isosteviol 2) influences on antituberculosis activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19345
Steviol, analytical standard
C20H30O3
H8664
Steviol hydrate, ≥98% (HPLC), powder
C20H30O3 · xH2O