EMAIL THIS PAGE TO A FRIEND
The Journal of antimicrobial chemotherapy
The enterococcal ABC transporter gene lsa(E) confers combined resistance to lincosamides, pleuromutilins and streptogramin A antibiotics in methicillin-susceptible and methicillin-resistant Staphylococcus aureus.
PMID 23047809
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0285
Pleuromutilin, ≥95%
C22H34O5