EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Association between aflatoxin M1 excreted in human urine samples with the consumption of milk and dairy products.
PMID 23052590
Abstract
This study aimed to find the association between urinary aflatoxin M(1) level and milk and dairy products consumption. Of 160 morning urine samples collected, aflatoxin M(1) was detected in 61.3 % samples (n = 98) [mean ± SD = 0.0234 ± 0.0177 ng/mL; range = 0-0.0747 ng/mL]. Of these positive samples, 67.3 % (n = 66) had levels above the limit of detection. Respondents with intake of milk and dairy products above median (67.79 g/day) had significantly high level of AFM(1) compared to those with low intake. A significant and positive association (φ = 0.286) was found between milk and dairy products consumption and urinary aflatoxin M(1) level.