EMAIL THIS PAGE TO A FRIEND
Parasitology research
Cytotoxic effect of acriflavine against clinical isolates of Acanthamoeba spp.
PMID 23052789
Abstract
Acanthamoeba keratitis (AK) is a potentially devastating and sight-threatening infection of the cornea caused by the ubiquitous free-living amoebae, Acanthamoeba species. Its eradication is difficult because the amoebas encyst, making it highly resistant to anti-amoebic drugs. Acriflavine neutral (ACF) has been used for treatment of microbial infections for humans and fishes. The aim of our study was to evaluate the time-dependent cytotoxicities of ACF against Acanthamoeba spp. Trophozoites and cysts of three different strains (strain PAT06 Acanthamoeba castellanii, strain 2HH Acanthamoeba hatchetti, and strain 11DS A. hatchetti) of Acanthamoeba spp. were tested. All strains had been isolated from patients suffering from a severe AK. The effects of the ACF with the concentrations ranging from 15 to 500 mg mL(-1) on the cytotoxicity of Acanthamoeba strains were examined. ACF showed a time- and dose-dependent amebicidal action on the trophozoites and cysts. Pat06 (A. castellanii) was the most resistant, while strain 11DS (A. hatchetti) was the most sensitive. As a result, ACF could be concluded as a new agent for the treatment of Acanthamoeba infections. On the other hand, it still needs to be further evaluated by in vivo test systems to confirm the efficiency of its biological effect.