EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
In Situ probing of newly synthesized peptidoglycan in live bacteria with fluorescent D-amino acids.
PMID 23055266
Abstract
Tracking a bug's life: Peptidoglycan (PG) of diverse bacteria is labeled by exploiting the tolerance of cells for incorporating different non-natural D-amino acids. These nontoxic D-amino acids preferably label the sites of active PG synthesis, thereby enabling fine spatiotemporal tracking of cell-wall dynamics in phylogenetically and morphologically diverse bacteria. HCC = 7-hydroxycoumarin, NBD = 7-nitrobenzofurazan, TAMRA = carboxytetramethylrhodamine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
650137
Benzofurazan, 97%
C6H4N2O