EMAIL THIS PAGE TO A FRIEND
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry
Immobilization of bacterial luciferase into poly(N-isopropylacrylamide) film for electrochemical control of a bioluminescence reaction.
PMID 23059999
Abstract
A poly(N-isopropylacrylamide) film was modified on an indium-tin oxide electrode in order to immobilize bacterial luciferase (BL) on the electrode surface. By using the modified electrode, flavin mononucleotide (FMN) was electrochemically reduced to FMNH(2), which is one of the substrates of the BL luminescence reaction, to control the bioluminescence reaction by BL. The BL reaction in the modified film could be promoted and controlled by the electrochemical generation of FMNH(2). This BL luminescence system was evaluated as a model system for the inhibitory assay of hydrophobic molecules on protein functions.