EMAIL THIS PAGE TO A FRIEND
Retina (Philadelphia, Pa.)
New tricks are not for everyone!
PMID 23060034
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
359238
Perfluorooctane, 98%
C8F18