EMAIL THIS PAGE TO A FRIEND
Journal of clinical gastroenterology
MiraLAX-Gatorade bowel prep versus GoLytely before screening colonoscopy: an endoscopic database study in a community hospital.
PMID 23060223
Abstract
Polyethylene glycol-3350 without electrolytes (MiraLAX; Schering-Plough Healthcare Products Inc.)+a carbohydrate-electrolyte solution (Gatorade; PepsiCo Inc.)+bisacodyl is frequently used for bowel cleansing, although limited data quantifies its efficacy and safety. No prior studies have assessed this in a community setting or with PM-only dosing, which is still used frequently. To compare the frequency of excellent/good/fair/poor bowel cleansing with PM-only dosing of MiraLAX-Gatorade-bisacodyl versus 4-liter GoLytely. This is a retrospective endoscopic database analysis of 50 years and older average-risk individuals with a normal screening colonoscopy at a community hospital and ambulatory endoscopy center. Data were extracted for the last 4 months when 4-liter GoLytely was the preferred bowel purgative and the first 4 months when 238-g MiraLAX in 64-ounce Gatorade and four 5-mg bisacodyl tablets became the preferred purgative. All patients used PM-only dosing of bowel purgative. A total of 778 subjects [GoLytely (n=395) vs. MiraLAX+Gatorade+bisacodyl (n=383)] were identified. Patients who took the MiraLAX bowel preparation were more likely to achieve an excellent/good bowel cleansing compared with patients taking the GoLytely preparation (93.3% vs. 89.3%, respectively; P=0.048). However, when only American Society of Anesthesiology (ASA) class I patients are studied, there was no difference in frequency of excellent/good bowel cleansing (91.1% vs 93.6%, respectively; P=0.498). No serious adverse events were identified. An excellent/good bowel cleansing was strongly associated with a recommendation for repeat colonoscopy in 10 years compared with patients with a fair cleansing (odds ratio=28.01; 95% confidence interval, 13.96-56.19). The MiraLAX+Gatorade+bisacodyl combination produces similar rates of excellent/good bowel cleansing as compared with GoLytely in most average-risk individuals undergoing colonoscopy for colorectal screening in a community setting.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1390
Bisacodyl, analytical standard, for drug analysis
C22H19NO4