EMAIL THIS PAGE TO A FRIEND
Acta poloniae pharmaceutica
Synthesis and in vitro antimicrobial activity of novel 2-(3-oxo-1,3-diarylpropylthio)acetic acid derivatives.
PMID 23061285
Abstract
A series of novel 2-(3-oxo-1,3-diarylpropylthio)acetic acid derivatives (3a-l) were prepared by base catalyzed addition of thioglycolic acid to chalcones (1a-l). The antibacterial activities of synthesized compounds were screened against human pathogenic microorganisms by employing the disk-diffusion technique. For the active compounds, also minimum inhibitory concentrations (MICs) were determined.