EMAIL THIS PAGE TO A FRIEND
Journal of neurosurgery
A randomized controlled study comparing omeprazole and cimetidine for the prophylaxis of stress-related upper gastrointestinal bleeding in patients with intracerebral hemorrhage.
PMID 23061387
Abstract
Patients with intracerebral hemorrhage (ICH) are at high risk for severe stress-related upper gastrointestinal (UGI) bleeding, which is predictive of higher mortality. The aim of this study was to evaluate the effectiveness of omeprazole and cimetidine compared with a placebo in the prevention and management of stress-related UGI bleeding in patients with ICH. In a single-center, randomized, placebo-controlled study, 184 surgically treated patients with CT-proven ICH within 72 hours of ictus and negative results for gastric occult blood testing were included. Of these patients, 165 who were qualified upon further evaluation were randomized into 3 groups: 58 patients received 40 mg intravenous omeprazole every 12 hours, 54 patients received 300 mg intravenous cimetidine every 6 hours, and 53 patients received a placebo. Patients whose gastric occult blood tests were positive at admission (n = 70) and during/after the prophylaxis procedure (n = 48) were treated with high-dose omeprazole at 80 mg bolus plus 8 mg/hr infusion for 3 days, followed by 40 mg intravenous omeprazole every 12 hours for 7 days. Of the 165 assessable patients, stress-related UGI bleeding occurred in 9 (15.5%) in the omeprazole group compared with 15 patients (27.8%) in the cimetidine group and 24 patients (45.3%) in the placebo group (p = 0.003). The occurrence of UGI bleeding was significantly related to death (p = 0.022). Nosocomial pneumonia occurred in 14 patients (24.1%) receiving omeprazole, 12 (22.2%) receiving cimetidine, and 8 (15.1%) receiving placebo (p > 0.05). In patients with UGI bleeding in which high-dose omeprazole was initiated, UGI bleeding arrested within the first 3 days in 103 patients (87.3%). Omeprazole significantly reduced the morbidity of stress-related UGI bleeding in patients with ICH due to its effective prophylactic effect without increasing the risk of nosocomial pneumonia, but it did not reduce the 1-month mortality or ICU stay. Further evaluation of high-dose omeprazole as the drug of choice for patients presenting with UGI bleeding is warranted. Clinical trial registration no.: ChiCTR-TRC-12001871, registered at the Chinese clinical trial registry (http://www.chictr.org/en/proj/show.aspx?proj=2384).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1089
Cimetidine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C10H16N6S · HCl