EMAIL THIS PAGE TO A FRIEND
Journal of veterinary internal medicine
Cervical myelomalacia in a lamb (Ovis aries) and an alpaca (Vicugna pacos) after attempted intramuscular injection.
PMID 23061655
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0261
Thiamphenicol
C12H15Cl2NO5S