EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Selective nanodecoration of modified cyclodextrin crystals with gold nanorods.
PMID 23062962
Abstract
Gold nanorods (AuNRs) stabilized by cetyltrimethylammonium bromide (CTAB) were deposited onto crystals of α-cyclodextrin (α-CD) inclusion compounds (ICs) that contained octanethiol (OT) as guest molecules. The nanodecoration was produced specifically at the {001} crystal planes through interaction between the -SH groups of the ICs and the AuNRs.