EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and biological evaluation of novel heterocyclic derivatives of combretastatin A-4.
PMID 23063401
Abstract
A novel series of combretastatin A-4 heterocyclic analogues was prepared by replacement of the B ring with indole, benzofurane or benzothiophene, attached at the C2 position. These compounds were evaluated for their abilities to inhibit tubulin assembly: derivative cis3b, having a benzothiophene, showed an activity similar to those of colchicine or deoxypodophyllotoxine. The antiproliferative and antimitotic properties of cis3b against keratinocyte cancer cell lines were also evaluated and the intracellular organization of microtubules in the cells after treatment with both stereoisomers of 3b was also determined, using confocal microscopy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B8002
2,3-Benzofuran, 99%
C8H6O