EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
The sesame lignan sesamin attenuates vascular dysfunction in streptozotocin diabetic rats: involvement of nitric oxide and oxidative stress.
PMID 23063541
Abstract
The effect of chronic administration of sesamin was studied on aortic reactivity of streptozotocin diabetic rats. Male diabetic rats received sesamin for 7 weeks after diabetes induction. Contractile responses to KCl and phenylephrine and relaxation response to acetylcholine were obtained from aortic rings. Maximum contractile response of endothelium-intact rings to phenylephrine was significantly lower in sesamin-treated diabetic rats relative to untreated diabetics and endothelium removal abolished this difference. Meanwhile, endothelium-dependent relaxation to acetylcholine was significantly higher in sesamin-treated diabetic rats as compared to diabetic ones and pretreatment of rings with nitric oxide synthase inhibitor N(G)-nitro-l-arginine methyl ester significantly attenuated the observed response. Two-month diabetes also resulted in an elevation of malondialdehyde and decreased superoxide dismutase activity and sesamin treatment significantly improved these changes. Therefore, chronic treatment of diabetic rats with sesamin could prevent some abnormal changes in vascular reactivity in diabetic rats through nitric oxide and via attenuation of oxidative stress and tissue integrity of endothelium is necessary for its beneficial effect.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S9314
Sesamin, ≥95%, crystalline
C20H18O6
59867
Sesamin, analytical standard
C20H18O6