EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
Short-term extractability of sulfadiazine after application to soils.
PMID 23063993
Abstract
The long-term environmental fate of the veterinary antibiotic sulfadiazine (SDZ) in soils is determined by a reversible sequestration into a residual fraction and an irreversible formation of non-extractable residues (NER), which can be described as first-order rate processes. However, the concentration dynamics of the resulting fractions of SDZ in soil show an unexplained rapid reduction of extractability during the first 24 h. We therefore investigated the short-term extractability of SDZ in two different soils under different SDZ application procedures over 24 h: with and without manure, for air-dried and for moist soils. In all batches, we observed an instantaneous loss of extractability on a time scale of minutes as well as kinetically determined sequestration and NER formation over 24 h. Data evaluation with a simple kinetic model led to the conclusion that application with manure accelerated the short-term formation of NER, whereas sequestration was very similar for all batches.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35033
Sulfadiazine, VETRANAL, analytical standard
C10H10N4O2S