EMAIL THIS PAGE TO A FRIEND
Molecular biology reports
In vitro inhibition of 7-ethoxyresorufin-O-deethylase (EROD) and p-nitrophenol hydroxylase (PNPH) activities by sesamin in hepatic microsomes from two fish species.
PMID 23065221
Abstract
In the present study, we investigated the effect of sesamin on CYP1A (7-ethoxyresorufin-O-deethylase, EROD) and CYP2E1-like activities (p-nitrophenol hydroxylase, PNPH) in hepatic microsomes obtained from Atlantic salmon (Salmo salar) and common carp (Cyprinus carpio). Addition of sesamin to the incubations in a concentration range from 1 to 200 μM decreased the activities of EROD and PNPH in a concentration dependent manner. It is likely that the inhibition of EROD was mechanism-based as demonstrated by the decrease in the IC50 value from 5.9 to 3.2 μM for A. salmon and from 7.9 to 3.0 μM for common carp when 5 min pre-incubation step was included. Similarly, PNPH activity was inhibited by sesamin with a decrease in the IC50 values from 61.7 to 15.2 μM for A. salmon and from 194.3 to 20.7 μM for common carp. Thus, our results indicated that sesamin can act as a mechanism-based inhibitor of EROD and PNPH activity with similar degree of inhibition in both fish species. More importantly, the inhibition of CYP1A, in addition to being mechanism-based, was competitive with K(i) value of 5.3 μM.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
59867
Sesamin, analytical standard
C20H18O6