EMAIL THIS PAGE TO A FRIEND
Biological trace element research
Aluminum induces osteoblast apoptosis through the oxidative stress-mediated JNK signaling pathway.
PMID 23065425
Abstract
Aluminum (Al) is considered to be a potentially toxic metal. Al exposure inhibits bone formation. Few studies have investigated the mechanism of inhibitory effects of Al on bone formation. Thus, in this study, osteoblasts were cultured and exposed to different concentrations of Al to investigate the mechanism behind the inhibitory effects of Al on bone formation. Al-treated osteoblasts showed signs of oxidative stress and a high apoptosis rate. The levels of osteoblasts activity markers (bone gamma-carboxyglutamic acid protein and bone alkaline phosphatase) were significantly lower in the Al-treated groups than in the control group. The c-Jun N-terminal kinase (JNK), a major signaling pathway in regulating cell apoptosis, was activated. The phosphorylation state of JNK was significantly increased. The mRNA and protein expression of c-Jun were both significantly upregulated. The pro-apoptotic genes (caspase 3, caspase 9, bax, and factor-related apoptosis ligand) were significantly increased. However, Bcl-2, an anti-apoptotic gene, was significantly decreased. In conclusion, the results of this study indicate that Al induces osteoblast apoptosis by activating the oxidative stress-mediated JNK pathway, which causes cell injuries and reduces the number and function of osteoblasts, thereby inhibiting bone formation.