EMAIL THIS PAGE TO A FRIEND
British journal of anaesthesia
Treatment of a life-threatening laryngeal bradykinin angio-oedema precipitated by dipeptidylpeptidase-4 inhibitor and angiotensin-I converting enzyme inhibitor with prothrombin complex concentrates.
PMID 23066003
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0094
Perindopril erbumine
C19H32N2O5 · C4H11N