EMAIL THIS PAGE TO A FRIEND
British journal of anaesthesia
Differential effect of phenylephrine and ephedrine on cerebral haemodynamics before carotid cross-clamping during carotid endarterectomy.
PMID 23066006
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
134910
(1R,2S)-(−)-Ephedrine, 98%
C10H15NO