EMAIL THIS PAGE TO A FRIEND
Psychiatry and clinical neurosciences
Quetiapine-induced insulin resistance after switching from blonanserin despite a loss in both bodyweight and waist circumference.
PMID 23066773
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B7188
Blonanserin, ≥98% (HPLC)
C23H30FN3