EMAIL THIS PAGE TO A FRIEND
Revista espanola de medicina nuclear e imagen molecular
Cerebral perfusion scintigraphy study as confirmation test of brain death in the process of organ donation for transplant.
PMID 23067532