EMAIL THIS PAGE TO A FRIEND
Acta biomaterialia
Moisture based three-dimensional printing of calcium phosphate structures for scaffold engineering.
PMID 23069318
Abstract
Powder based three-dimensional printing (3DP) allows great versatility in material and geometry. These characteristics make 3DP an interesting method for the production of tissue engineering scaffolds. However, 3DP has major limitations, such as limited resolution and accuracy, hence preventing the widespread application of this method within scaffold engineering [corrected].In order to reduce these limitations deeper understanding of the complex interactions between powder, binder and roller during 3DP is needed. In the past a lot of effort has been invested to optimize the powder properties for 3DP for a certain layer thickness. Using a powder optimized for an 88 μm layer thickness, this study systematically quantifies the surface roughness and geometrical accuracy in printed specimens and assesses their variation upon changes of different critical parameters such as the moisture application time (0, 5, 10 and 20s), layer thickness (44 and 88 μm) and the number of specimens printed per batch (6 and 12). A best surface roughness value of 25 μm was measured with a moisture application time (using a custom made moisture application device mounted on a linear stage carrying the print head) of 5s and a layer thickness of 44 μm. Geometrical accuracy was generally higher for the 88 μm thick layer, due to a less critical powder bed stability. Moisture application enabled 3DP of a 44 μm thick layer and improved the accuracy even for a powder initially optimized for 88 μm. Moreover, recycling of the humidified powder was not only possible but, in terms of reactivity, even beneficial. In conclusion, moisture-based 3DP is a promising approach for high resolution 3DP of scaffolds.