EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Association between DPP6 polymorphism and the risk of progressive multiple sclerosis in Northern and Southern Europeans.
PMID 23069673
Abstract
In this study, we investigated the role of the dipeptidyl-peptidase-6 (DPP6) gene in the etiopathogenesis of progressive forms of multiple sclerosis (PrMS). This gene emerged as a candidate gene in a genome-wide association study (GWAS) performed in an Italian sample of PrMS and controls in which two SNPs located in the gene (rs6956703 and rs11767658) showed evidence of association (nominal p-value<10(-4)) (Martinelli-Boneschi et al.) [18]. Moreover, the gene is highly expressed in the central nervous system, and it has been found to be associated with sporadic cases of amyotrophic lateral sclerosis which shares some feature with PrMS. We genotyped 19 SNPs selected using a direct and tagging approach in 244 Italian PrMS and 225 controls, and we measured the expression levels of the gene in 13 PrMS cases and 25 controls. Five out of 19 SNPs were found to be associated with the disease (adjusted p<0.05), and they have been tested in an independent sample of 179 primary progressive MS and 198 controls from Northern Europe. None of the SNPs was replicated, but combined analysis confirmed the presence of association for rs2046748 (p=2.5×10(-3),OR=1.82, 95%CI=1.24-2.69). These results, inflated by the limited sample size determined by the rarity of this condition, suggest a possible role of this gene in the susceptibility to PrMS, at least in Southern Europeans. Moreover, DPP6 was over-expressed in PrMS patients compared to controls.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3321 Dipeptidyl Peptidase VII human, recombinant, expressed in Sf9 cells