EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Synthesis of sterically encumbered C10-arylated benzo[h]quinolines using ortho-substituted aryl boronic acids.
PMID 23069777
Abstract
The challenging coupling of 10-halobenzo[h]quinolines with ortho-substituted aryl boronic acids has been achieved using Pd(OAc)(2)/P(O)Ph(3) as the catalytic system. High yields were obtained for diversely functionalised substrates under mild reaction conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123617
Benzo[h]quinoline, 97%
C13H9N