EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Volumetric stability of lipid bilayers.
PMID 23069984
Abstract
In agreement with recent reports, a commercial densimeter has yielded a gradual decrease in lipid molecular volume of DPPC multilamellar vesicle dispersions in the gel phase upon repeated thermal cycling between 10 °C and 50 °C. The considerable size of this decrease would have significant implications for the physical chemistry of biomembranes. In contrast, neutral buoyancy measurements performed with similar thermal cycling indicate no gradual change in lipid volume in the gel phase at 20 °C. Remixing the lipid in the densimeter shows that the apparent volume decrease is an artifact. We conclude that gel phase DPPC bilayers exist in a volumetrically stable phase.